2-(4-Chlorophenoxy)malondialdehyde 95+%

Lieferant: Apollo Scientific
Achtung

OR7635-1G
APOSOR7635-1GEA 63.8 EUR
APOSOR7635-1G
2-(4-Chlorophenoxy)malondialdehyde 95+%
2-(4-Chlorophenoxy)malondialdehyde
Formel: C₉H₇ClO₃
Molecular Weight: 198,61 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426063
CAS-Nummer: 849021-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR