1,2-Dimethyl-1H-indole-3-carboxaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

OR7637-1G OR7637-250MG
APOSOR7637-1GEA 29 EUR
APOSOR7637-1G APOSOR7637-250MG
1,2-Dimethyl-1H-indole-3-carboxaldehyde 95+%
1,2-Dimethyl-1H-indole-3-carboxaldehyde
Formel: C₁₁H₁₁NO
Schmelzpunkt: 127…129 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00745294
CAS-Nummer: 38292-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR