2-(3-Methoxyphenyl)malondialdehyde 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7645-1G
APOSOR7645-1GEA
300
EUR
APOSOR7645-1G
2-(3-Methoxyphenyl)malondialdehyde 95+%
2-(3-Methoxyphenyl)malondialdehyde
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178,19 g/mol Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02261923 CAS-Nummer: 857500-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...