2-Amino-4-phenylpyrimidine

Lieferant: Apollo Scientific

OR7650-1G
APOSOR7650-1GEA 72.5 EUR
APOSOR7650-1G
2-Amino-4-phenylpyrimidine
2-Amino-4-phenylpyrimidine
Formel: C₁₀H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00094706
CAS-Nummer: 2305-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR