2'-Chloracetanilid 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2-Chlorphenyl)acetamid

OR7655-5G
APOSOR7655-5GEA 21.9 EUR
APOSOR7655-5G
2'-Chloracetanilid 98+%
2'-Chloracetanilid
Formel: C₈H₈ClNO
Molecular Weight: 169,61 g/mol
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045169
CAS-Nummer: 533-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR