4-Phenyl[1,3,5]triazine-2-thiol

Lieferant: Apollo Scientific
Achtung

OR7661-1G
APOSOR7661-1GEA 173 EUR
APOSOR7661-1G
4-Phenyl[1,3,5]triazine-2-thiol
4-Phenyl[1,3,5]triazine-2-thiol
Formel: C₉H₇N₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09702439
CAS-Nummer: 1000018-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR