1-Benzyl-1,4-diazepan-5-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzylhomopiperazin-5-one

OR7666-1G
APOSOR7666-1GEA 23.2 EUR
APOSOR7666-1G
1-Benzyl-1,4-diazepan-5-one
1-Benzyl-1,4-diazepan-5-one
Formel: C₁₂H₁₆N₂O
Molecular Weight: 204,2700 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00225259
CAS-Nummer: 55186-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR