2,4-Dibenzyloxy-5-pyrimidinboronsäure

Lieferant: Apollo Scientific
Achtung

OR7669-1G
APOSOR7669-1GEA 183 EUR
APOSOR7669-1G
2,4-Dibenzyloxy-5-pyrimidinboronsäure
2,4-Dibenzyloxy-5-pyrimidinboronsäure
Formel: C₁₈H₁₇BN₂O₄
Lagertemperatur: Kühlschrank
MDL: MFCD01318938
CAS-Nummer: 70523-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR