5-(3-Chlorphenyl)oxazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR7671-100MG
OR7671-1G
OR7671-250MG
APOSOR7671-100MGEA
0
EUR
APOSOR7671-100MG
APOSOR7671-1G
APOSOR7671-250MG
5-(3-Chlorphenyl)oxazol 98%
5-(3-Chlorophenyl)oxazole
Formel:
C₉H₆ClNO Schmelzpunkt: 46…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084974 CAS-Nummer: 89808-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...