3,5-Bis(methylsulphonyl)aniline 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7687-250MG
APOSOR7687-250MGEA
58
EUR
APOSOR7687-250MG
3,5-Bis(methylsulphonyl)aniline 95+%
3,5-Bis(methylsulphonyl)aniline
Formel:
C₈H₁₁NO₄S₂ Schmelzpunkt: 202…205 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01464121 CAS-Nummer: 51859-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...