3-Methylbenzo[b]thiophen-2-carbaldehyd 97%

Lieferant: Apollo Scientific

OR7692-10G OR7692-25G OR7692-5G
APOSOR7692-10GEA 408 EUR
APOSOR7692-10G APOSOR7692-25G APOSOR7692-5G
3-Methylbenzo[b]thiophen-2-carbaldehyd 97%
3-Methylbenzo[b]thiophen-2-carbaldehyd
Formel: C₁₀H₈OS
Molecular Weight: 176.24 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052307
CAS-Nummer: 22053-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR