3-(Benzyloxy)propane-1,2-diol 95+%

Lieferant: Apollo Scientific

OR7701T-25G
APOSOR7701T-25GEA 94.4 EUR
APOSOR7701T-25G
3-(Benzyloxy)propane-1,2-diol 95+%
3-(Benzyloxy)propane-1,2-diol
Formel: C₁₀H₁₄O₃
Siedepunkt: 140…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016633
CAS-Nummer: 4799-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR