tert-Butyl-3-(aminomethyl)phenylcarbamate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminomethyl)aniline, 1-BOC protected, 3-(Aminomethyl)-1-N-Boc-aniline

OR7705-1G
APOSOR7705-1GEA 50.9 EUR
APOSOR7705-1G
tert-Butyl-3-(aminomethyl)phenylcarbamate
tert-Butyl-3-(aminomethyl)phenylcarbamate
Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,29 g/mol
Siedepunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01317790
CAS-Nummer: 205318-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR