2-tert-Butyl-1H-indol-5-amine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-2-(tert-butyl)-1H-indole
OR7706-1G
APOSOR7706-1GEA
203
EUR
APOSOR7706-1G
2-tert-Butyl-1H-indol-5-amine 95+%
2-tert-Butyl-1H-indol-5-amine
Formel:
C₁₂H₁₆N₂ Molecular Weight: 188,27 g/mol Schmelzpunkt: 151…154 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD04037904 CAS-Nummer: 682357-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...