2-(Chloromethyl)-4,5-dihydro-1H-imidazole hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR7709-5G OR7709-100G OR7709-25G
APOSOR7709-5GEA 29 EUR
APOSOR7709-5G APOSOR7709-100G APOSOR7709-25G
2-(Chloromethyl)-4,5-dihydro-1H-imidazole hydrochloride 95%
2-(Chloromethyl)-4,5-dihydro-1H-imidazole hydrochloride
Formel: C₄H₈Cl₂N₂
Schmelzpunkt: 195…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115823
CAS-Nummer: 13338-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR