1-[2-Chloro-4-methyl-5-(methylsulphonyl)phenyl]piperazine
Lieferant: Apollo Scientific
|
Achtung
|
OR7711-1G
APOSOR7711-1GEA
112
EUR
APOSOR7711-1G
1-[2-Chloro-4-methyl-5-(methylsulphonyl)phenyl]piperazine
1-[2-Chloro-4-methyl-5-(methylsulphonyl)phenyl]piperazine
Formel:
C₁₂H₁₇ClN₂O₂S Schmelzpunkt: 158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037928 CAS-Nummer: 849035-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...