1-(2,6-Dimethoxybenzyl)-N-methylpiperidine-4-amine 95+%

Lieferant: Apollo Scientific

OR7725-1G
APOSOR7725-1GEA 317 EUR
APOSOR7725-1G
1-(2,6-Dimethoxybenzyl)-N-methylpiperidine-4-amine 95+%
1-(2,6-Dimethoxybenzyl)-N-methylpiperidine-4-amine
Formel: C₁₅H₂₄N₂O₂
Molecular Weight: 264,37 g/mol
Schmelzpunkt: 58…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319290
CAS-Nummer: 397245-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR