3-Chlor-1H-indazol 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Chlor-1H-indazol

OR7740-5G
APOSOR7740-5GEA 121 EUR
APOSOR7740-5G
3-Chlor-1H-indazol 95+%
3-Chloro-1H-indazole
Formel: C₇H₅ClN₂
Molecular Weight: 152,58 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067527
CAS-Nummer: 29110-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR