9,10-Anthrachinon-2-carbonsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Anthraquinone-2-carboxylic acid, 9,10-Dioxo-9,10-dihydroanthracene-2-carboxylic acid

OR7750-5G
APOSOR7750-5GEA 43.5 EUR
APOSOR7750-5G
9,10-Anthrachinon-2-carbonsäure 95%
9,10-Anthrachinon-2-carbonsäure
Formel: C₁₅H₈O₄
Molecular Weight: 252.22 g/mol
Schmelzpunkt: 287…289 °C
MDL: MFCD00001231
CAS-Nummer: 117-78-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR