4-Hydrazinobenzoesäure

Lieferant: Apollo Scientific
Achtung

OR7755-10G OR7755-100G OR7755-25G
APOSOR7755-10GEA 16.1 EUR
APOSOR7755-10G APOSOR7755-100G APOSOR7755-25G
4-Hydrazinobenzoesäure
4-Hydrazinobenzoic acid
Formel: C₇H₈N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD00007581
CAS-Nummer: 619-67-0
EINECS: 210-609-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR