4-Hydrazinobenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7755-10G
OR7755-100G
OR7755-25G
APOSOR7755-10GEA
16.1
EUR
APOSOR7755-10G
APOSOR7755-100G
APOSOR7755-25G
4-Hydrazinobenzoesäure
4-Hydrazinobenzoic acid
Formel:
C₇H₈N₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD00007581 CAS-Nummer: 619-67-0 EINECS: 210-609-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...