3,5-Dimethyl-4-iodisoxazol

Lieferant: Apollo Scientific

OR7771-100G
APOSOR7771-100GEA 91.5 EUR
APOSOR7771-100G
3,5-Dimethyl-4-iodisoxazol
3,5-Dimethyl-4-iodoisoxazole
Formel: C₅H₆INO
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173743
CAS-Nummer: 10557-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR