N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Aminomethyl)acetanilide hydrochloride
OR7774-1G
OR7774-250MG
APOSOR7774-1GEA
126
EUR
APOSOR7774-1G
APOSOR7774-250MG
N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%
N-[3-Aminomethyl)phenyl]acetamide hydrochloride
Formel:
C₉H₁₃ClN₂O Schmelzpunkt: 230…233 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039171 CAS-Nummer: 238428-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...