N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminomethyl)acetanilide hydrochloride

OR7774-1G OR7774-250MG
APOSOR7774-1GEA 126 EUR
APOSOR7774-1G APOSOR7774-250MG
N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%
N-[3-Aminomethyl)phenyl]acetamide hydrochloride
Formel: C₉H₁₃ClN₂O
Schmelzpunkt: 230…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039171
CAS-Nummer: 238428-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR