5-Bromo-1,3-dimethyluracil 95+%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine, 5-Bromo-1,3-dimethylpyrimidine-2,4(1H,3H)-dione

OR7780-1G
APOSOR7780-1GEA 71.3 EUR
APOSOR7780-1G
5-Bromo-1,3-dimethyluracil 95+%
5-Bromo-1,3-dimethyluracil
Formel: C₆H₇BrN₂O₂
Molecular Weight: 219,036 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191846
CAS-Nummer: 7033-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR