1-[(2-Methoxyethyl)amino]-1-(4-methylsulphonylphenyl)ethane 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(2-Methoxyethyl)-alpha-methyl-4-(methylsulphonyl)benzylamine

OR7787-1G
APOSOR7787-1GEA 84.1 EUR
APOSOR7787-1G
1-[(2-Methoxyethyl)amino]-1-(4-methylsulphonylphenyl)ethane 95+%
1-[(2-Methoxyethyl)amino]-1-(4-methylsulphonylphenyl)ethane
Formel: C₁₂H₁₉NO₃S
Molecular Weight: 257,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00203232
CAS-Nummer: 849924-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR