1-[4-(Methylsulphonyl)-2-nitrophenyl]-1,4-diazepane 95+%

Lieferant: Apollo Scientific

Synonyme: 1-[4-(Methylsulphonyl)-2-nitrophenyl]homopiperazine

OR7795-1G
APOSOR7795-1GEA 285 EUR
APOSOR7795-1G
1-[4-(Methylsulphonyl)-2-nitrophenyl]-1,4-diazepane 95+%
1-[4-(Methylsulphonyl)-2-nitrophenyl]-1,4-diazepane
Formel: C₁₂H₁₇N₃O₄S
Molecular Weight: 299,35 g/mol
Schmelzpunkt: 136…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037963
CAS-Nummer: 849035-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR