2-Bromo-5-methoxy-N,N-dimethylbenzylamine

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-3-[(dimethylamino)methyl]anisole, (2-Bromo-5-methoxyphenyl)-N,N-dimethylmethylamine

OR7801-1G
APOSOR7801-1GEA 58.2 EUR
APOSOR7801-1G
2-Bromo-5-methoxy-N,N-dimethylbenzylamine
2-Bromo-5-methoxy-N,N-dimethylbenzylamine
Formel: C₁₀H₁₄BrNO
Molecular Weight: 244,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657799
CAS-Nummer: 10126-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR