4-[(Benzyloxy)methyl]-1,3-dioxolan-2-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzylglycerol-2,3-carbonate, 4-[(Benzyloxy)methyl]-2-oxo-1,3-dioxolane

OR7801T-1G
APOSOR7801T-1GEA 139 EUR
APOSOR7801T-1G
4-[(Benzyloxy)methyl]-1,3-dioxolan-2-one
4-[(Benzyloxy)methyl]-1,3-dioxolan-2-one
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,211 g/mol
Siedepunkt: 165 °C (0,8 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD00269930
CAS-Nummer: 949-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR