4-(4-Nitrophenyl)-1H-pyrazole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7805-1G
OR7805-5G
OR7805-25G
APOSOR7805-1GEA
142
EUR
APOSOR7805-1G
APOSOR7805-5G
APOSOR7805-25G
4-(4-Nitrophenyl)-1H-pyrazole 95+%
4-(4-Nitrophenyl)-1H-pyrazole
Formel:
C₉H₇N₃O₂ Schmelzpunkt: 205…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037972 CAS-Nummer: 114474-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...