2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[3-(1,4-Diazin-2-yl)-1,2,4-oxadiazol-5-yl]piperidine, 5-(Piperidin-4-yl)-3-(pyrazin-2-yl)-1,2,4-oxadiazole

OR7807-250MG
APOSOR7807-250MGEA 92.8 EUR
APOSOR7807-250MG
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine 95+%
2-[5-(Piperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyrazine
Formel: C₁₁H₁₃N₅O
Molecular Weight: 231,254 g/mol
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426077
CAS-Nummer: 849925-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR