4-[3-(Thien-2-yl)[1,2,4]oxadiazol-5-yl]piperidine 95+%

Lieferant: Apollo Scientific
Achtung

OR7813-100MG
APOSOR7813-100MGEA 21.9 EUR
APOSOR7813-100MG
4-[3-(Thien-2-yl)[1,2,4]oxadiazol-5-yl]piperidine 95+%
4-[3-(Thien-2-yl)[1,2,4]oxadiazol-5-yl]piperidine
Formel: C₁₁H₁₃N₃OS
Molecular Weight: 235,31 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426076
CAS-Nummer: 244272-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR