4-(4-Benzyl-1,4-diazepan-1-yl)-4-oxobutanoic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Benzylhomopiperazin-1-yl)-4-oxobutanoic acid

OR7821-1G
APOSOR7821-1GEA 235 EUR
APOSOR7821-1G
4-(4-Benzyl-1,4-diazepan-1-yl)-4-oxobutanoic acid 95+%
4-(4-Benzyl-1,4-diazepan-1-yl)-4-oxobutanoic acid
Formel: C₁₆H₂₂N₂O₃
Molecular Weight: 290,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800863
CAS-Nummer: 396105-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR