4-Brom-2-(methylsulfonyl)benzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7823-1G
APOSOR7823-1GEA
55.1
EUR
APOSOR7823-1G
4-Brom-2-(methylsulfonyl)benzoesäure
4-Bromo-2-(methylsulphonyl)benzoic acid
Formel:
C₈H₇BrO₄S Schmelzpunkt: 220…224 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037926 CAS-Nummer: 142994-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...