1-(5-Bromopyrimidin-2-yl)piperidine-4-carboxylic acid 95+%

Lieferant: Apollo Scientific

OR7826-1G
APOSOR7826-1GEA 169 EUR
APOSOR7826-1G
1-(5-Bromopyrimidin-2-yl)piperidine-4-carboxylic acid 95+%
1-(5-Bromopyrimidin-2-yl)piperidine-4-carboxylic acid
Formel: C₁₀H₁₂BrN₃O₂
Molecular Weight: 286.14 g/mol
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426070
CAS-Nummer: 799283-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR