3-(tert-Butoxycarbonyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR7828-1G OR7828-5G OR7828-10G
APOSOR7828-1GEA 84.1 EUR
APOSOR7828-1G APOSOR7828-5G APOSOR7828-10G
3-(tert-Butoxycarbonyl)benzoic acid
3-(tert-Butoxycarbonyl)benzoic acid
Formel: C₁₂H₁₄O₄
Schmelzpunkt: 152…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037899
CAS-Nummer: 33704-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR