3-(tert-Butoxycarbonyl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR7828-1G
OR7828-5G
OR7828-10G
APOSOR7828-1GEA
84.1
EUR
APOSOR7828-1G
APOSOR7828-5G
APOSOR7828-10G
3-(tert-Butoxycarbonyl)benzoic acid
3-(tert-Butoxycarbonyl)benzoic acid
Formel:
C₁₂H₁₄O₄ Schmelzpunkt: 152…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037899 CAS-Nummer: 33704-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...