4-Chlor-2-(methylsulfonyl)benzoesäure 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7834-250MG
APOSOR7834-250MGEA
0
EUR
APOSOR7834-250MG
4-Chlor-2-(methylsulfonyl)benzoesäure 95+%
4-Chloro-2-(methylsulphonyl)benzoic acid
Formel:
C₈H₇ClO₄S Schmelzpunkt: 194…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037924 CAS-Nummer: 142994-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...