3-(2,3-Dihydrobenzofuran-5-yl)propanoic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-1-benzofuran-5-propanoic acid
OR7838-250MG
APOSOR7838-250MGEA
59.6
EUR
APOSOR7838-250MG
3-(2,3-Dihydrobenzofuran-5-yl)propanoic acid 95+%
3-(2,3-Dihydrobenzofuran-5-yl)propanoic acid
Formel:
C₁₁H₁₂O₃ Molecular Weight: 192,22 g/mol Schmelzpunkt: 144…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03102298 CAS-Nummer: 215057-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...