4-(8,9-Dihydropyrido[2,3-b]-1,6-naphthyridin-7(6H)-yl)-4-oxobutanoic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(7,8-Dihydro-5H-1,6,9-triazaanthracen-6-yl)-4-oxobutanoic acid

OR7842-1G
APOSOR7842-1GEA 294 EUR
APOSOR7842-1G
4-(8,9-Dihydropyrido[2,3-b]-1,6-naphthyridin-7(6H)-yl)-4-oxobutanoic acid 95+%
4-(8,9-Dihydropyrido[2,3-b]-1,6-naphthyridin-7(6H)-yl)-4-oxobutanoic acid
Formel: C₁₅H₁₅N₃O₃
Molecular Weight: 285,31 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203038
CAS-Nummer: 389117-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR