Benzylcarbazat

Lieferant: Apollo Scientific
Gefahr

Synonyme: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate

OR7846-500G
APOSOR7846-500GEA 128 EUR
APOSOR7846-500G
Benzylcarbazat
Benzylcarbazat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 65…70 °C
Flammpunkt: 110 °C
MDL: MFCD00041890
CAS-Nummer: 5331-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR