1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR7849-1G
APOSOR7849-1GEA 58 EUR
APOSOR7849-1G
1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid
1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid
Formel: C₁₂H₁₇N₃O₂
Molecular Weight: 235.29 g/mol
Schmelzpunkt: 158…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426079
CAS-Nummer: 797028-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR