6-(Benzyloxy)-7H-purin-2-amin
Lieferant: Apollo Scientific
Synonyme:
O6-Benzylguanine
OR7851T-50MG
APOSOR7851T-50MGEA
118
EUR
APOSOR7851T-50MG
6-(Benzyloxy)-7H-purin-2-amin
6-(Benzyloxy)-7H-purin-2-amine
An effective substrate & efficient inactivator of DNA alkyl transferase. Protects against carcinogenic & therapeutic alkylating agents.
Formel:
C12H11N5O Molecular Weight: 241,2 g/mol Schmelzpunkt: 193 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269931 CAS-Nummer: 19916-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...