3-Brom-4-chlorbenzoesäure

Lieferant: Apollo Scientific
Achtung

OR7862-5G
APOSOR7862-5GEA 21.9 EUR
APOSOR7862-5G
3-Brom-4-chlorbenzoesäure
3-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
Schmelzpunkt: 218…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079706
CAS-Nummer: 42860-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR