4-(1,7-Naphthyridin-8-ylamino)-4-oxobutanoic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR7866-1G
APOSOR7866-1GEA 108 EUR
APOSOR7866-1G
4-(1,7-Naphthyridin-8-ylamino)-4-oxobutanoic acid 95+%
4-(1,7-Naphthyridin-8-ylamino)-4-oxobutanoic acid
Formel: C₁₂H₁₁N₃O₃
Schmelzpunkt: 66…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319225
CAS-Nummer: 396682-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR