2-(1,6-Naphthyridin-2-yl)benzoic acid hydrate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Carboxyphenyl)-1,6-naphthyridine hydrate

OR7867-500MG
APOSOR7867-500MGEA 63.8 EUR
APOSOR7867-500MG
2-(1,6-Naphthyridin-2-yl)benzoic acid hydrate 95+%
2-(1,6-Naphthyridin-2-yl)benzoic acid hydrate
Formel: C₁₅H₁₀N₂O₂·H₂O
Schmelzpunkt: 175…185 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD08445563
CAS-Nummer: 1172902-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR