3-(4-Chlorphenyl)propionsäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Chlorphenyl)propansäure

OR7880-25G
APOSOR7880-25GEA 94.4 EUR
APOSOR7880-25G
3-(4-Chlorphenyl)propionsäure 98%
3-(4-Chlorphenyl)propionsäure
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Schmelzpunkt: 127…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016555
CAS-Nummer: 2019-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR