5-Bromo-2-hydrazinopyrimidine

Lieferant: Apollo Scientific
Achtung

OR7905-1G
APOSOR7905-1GEA 21.9 EUR
APOSOR7905-1G
5-Bromo-2-hydrazinopyrimidine
5-Bromo-2-hydrazinopyrimidine
Formel: C₄H₅BrN₄
Molecular Weight: 189.0100 g/mol
Schmelzpunkt: 205…207 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD03452780
CAS-Nummer: 823-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR