Methyl-1,6-naphthyridine-2-carboxylate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1,6-naphthyridine-2-carboxylate

OR7910-1G
APOSOR7910-1GEA 261 EUR
APOSOR7910-1G
Methyl-1,6-naphthyridine-2-carboxylate 95+%
Methyl-1,6-naphthyridine-2-carboxylate
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188.19 g/mol
Schmelzpunkt: 119…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202909
CAS-Nummer: 338760-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR