Methyl-3-(aminomethyl)benzoate hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-(aminomethyl)benzoate hydrochloride
OR7914-1G
OR7914-5G
OR7914-25G
APOSOR7914-1GEA
29
EUR
APOSOR7914-1G
APOSOR7914-5G
APOSOR7914-25G
Methyl-3-(aminomethyl)benzoate hydrochloride
Methyl-3-(aminomethyl)benzoate hydrochloride
Formel:
C₉H₁₂ClNO₂ Schmelzpunkt: 179…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039173 CAS-Nummer: 17841-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...